Sand, quartz, dried, washed and dried
Supplier: SPECTRUM CHEMICAL MFG CORP
S1020-2.5KG
S1020-500GM
700000-334EA
535
USD
700000-334
700000-338
Sand, quartz, dried, washed and dried
Sand, quartz
Sand, Washed and Dried is also known as a textural class of soil that contains more than 85% of sand particles. It is a granuar material made up of mineral and rock particles.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
SiO₂ Boiling Pt: 2230 °C (1013 hPa) Melting Pt: 1427 °C Density: 2.622…2.645 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011232 CAS Number: 14808-60-7 EINECS: 238-878-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...