L(+)-Rhamnose monohydrate

Synonyms: 6-Deoxy-L(+)-mannose monohydrate(2R,3R,4S,5S)-2,3,4,5-tetrahydroxyhexanal hydrate

RH105-25GM RH105-5KGBL RH105-5GM RH105-100GM RH105-04
700009-704EA 69.64 USD
700009-704 700009-706 CA11024-406 700009-700 700009-702
L(+)-Rhamnose monohydrate
L(+)-Rhamnose monohydrate

L-Rhamnose, Monohydrate, also known as Isodulcit, is a naturally occurring deoxy sugar.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₆H₁₂O₅·1H₂O
MW: 182.17 g/mol
Melting Pt: 85…95 °C
Density: 1.47 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00071591
CAS Number: 10030-85-0
EINECS: 222-793-4
Merck Index: 12,08338

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR