Prednisolone, anhydrous ≥97%, micronized USP
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Prednisolonum
PR101-1KGBL
95035-302EA
10851
USD
95035-302
Prednisolone, anhydrous ≥97%, micronized USP
Prednisolone
Prednisolone, Anhydrous, Micronized, USP is a steroid medication used to treat certain types of allergies, inflammatory conditions, autoimmune disorders, and cancers. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities
Formula:
C₂₁H₂₈O₅ MW: 360.45 g/mol Boiling Pt: 571 °C (1013 hPa) Melting Pt: 246 °C Density: 1.212 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003649 CAS Number: 50-24-8 EINECS: 200-021-7 |
Specification Test Results
Assay | 97% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...