Potassium carbonate sesquihydrate ≥99%, crystals FCC
|
Warning
|
Potassium Carbonate, Sesquihydrate, Crystal, FCC is used as a meat tenderizer, a source of dietary potassium and a buffering agent in wine production. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formula:
CK₂O₃·1.5H₂O MW: 165.23 g/mol Melting Pt: 891 °C Density: 2.29 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149925 CAS Number: 6381-79-9 EINECS: 209-529-3 |
Specification Test Results
Assay | 99% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...