Potassium benzoate ≥99% FCC
Supplier: SPECTRUM CHEMICAL MFG CORP
Potassium Benzoate, FCC is used as a preservative that limits the growth of yeast, mold and some bacteria. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formula:
C₇H₅KO₂ MW: 160.21 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00013061 CAS Number: 582-25-2 EINECS: 209-481-3 |
Specification Test Results
Assay | 99% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...