Piperazine hexahydrate ≥98%

Danger

Synonyms: Piperazine hydrate

PI122-1KG PI122-100GM
700008-816EA 151.68 USD
700008-816 700008-812
Piperazine hexahydrate ≥98%
Piperazine hexahydrate

Piperazine, Hexahydrate, also known as Diethylenediamine, exists as small alkaline deliquescent crystals with a saline taste. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Formula: C₄H₁₀N₂·6H₂O
MW: 194.23 g/mol
Boiling Pt: 145 °C (1013 hPa)
Melting Pt: 44…45 °C
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00149389
CAS Number: 142-63-2
EINECS: 203-808-3
UN: 2579
ADR: 8,III
Merck Index: 12,07617

Order Now

Specification Test Results

Assay 98% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR