Polyvinylacetate, M.W. ∼ 50,000

Synonyms: PVA

P1797-25GM P1797-2.5KG P1797-500GM
700012-226EA 1102.15 USD
700012-226 700012-228 700012-230
Polyvinylacetate, M.W. ∼ 50,000
Polyvinylacetate

Polyvinyl Acetate, also known as PVA, is a synthetic polymer used in textiles, papermaking, and several types of coatings.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: [CH₂CH(O₂CCH₃)]n
Storage Temperature: Ambient
MDL Number: MFCD00084457
CAS Number: 9003-20-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR