Palmitic acid FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Hexadecanoic acidHexadecylic acid

P1513-500GM P1513-2.5KG P1513-50KGBL
700005-020EA 645 USD
700005-020 700005-022 700005-024
Palmitic acid FCC
Palmitic acid

Palmitic Acid, FCC adds texture and "mouth feel" to processed foods. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₁₆H₃₂O₂
MW: 256.43 g/mol
Boiling Pt: 352 °C (1013 hPa)
Melting Pt: 61…63 °C
Density: 0.853 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002747
CAS Number: 57-10-3
EINECS: 200-312-9
Merck Index: 12,07128

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR