Methyl oleate

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: cis-Oleic Acid Methyl EsterOleic acid methyl ester

O2097-1GM O2097-25GM O2097-100GM O2097-5GM
700011-834EA 266 USD
700011-834 700011-836 700011-830 700011-832
Methyl oleate
Methyl oleate

cis-Oleic Acid Methyl Ester is a fatty acid methyl ester intermediate for detergents, emulsifiers, wetting agents, stabilizers, textile treatment, plasticizers for duplicating inks, rubbers, and waxes.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₉H₃₆O₂
MW: 296.49 g/mol
Boiling Pt: ∼218.5 °C (1013 hPa)
Melting Pt: –20 °C
Density: 0.879
Storage Temperature: Refrigerator
MDL Number: MFCD00009578
CAS Number: 112-62-9
EINECS: 203-992-5
Merck Index: 12,06965

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR