1-Octanol ≥99% ACS
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Warning
|
Synonyms:
n-Octanol
O1004-20LT
O1004-500MLG
O1004-4LTGL
O1004-200LTB
700002-694EA
4006
USD
700002-694
700002-700
700002-698
700002-696
1-Octanol ≥99% ACS
1-Octanol
1-Octanol, Reagent, ACS is a fatty alcohol that functions as a fragrance ingredient. It is a liquid and is insoluble in cold water. As an ACS grade reagent, it is primarily used as a solvent. Spectrum Chemical manufactured ACS compounds are used as the quality standards against which other substances are graded and have met the toughest regulatory standards for quality and pureness
Formula:
H₃C(CH₂)₆CH₂OH MW: 130.23 g/mol Boiling Pt: 195 °C (1013 hPa) Melting Pt: –16 °C Density: 0.827 Flash Pt: 81 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002988 CAS Number: 111-87-5 EINECS: 203-917-6 Merck Index: 12,06849 |
Specification Test Results
Assay | 99% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...