(±)-2-Nonanol ≥99%

Synonyms: (±)-2-Hydroxy nonane(±)-Heptyl methyl carbinol(±)-Methyl heptyl carbinol

N2364-25GM N2364-100GM N2364-5GM
700002-800EA 50.56 USD
700002-800 700002-798 700002-796
(±)-2-Nonanol ≥99%
(±)-2-Nonanol

2-Nonanol is a flavor and fragrant ingredient.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₉H₂₀O
MW: 144.26 g/mol
Boiling Pt: 193 °C (1013 hPa)
Melting Pt: –35 °C
Density: 0.827
Storage Temperature: Ambient
MDL Number: MFCD00004593
CAS Number: 628-99-9
EINECS: 211-065-1

Order Now

Specification Test Results

Assay 99% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR