β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na₂, reduced form), reduced

Synonyms: ((2R,3S,4R,5R)-5-(6-Aminopurin-9-yl)-3,4-dihydroxy-oxolan-2-yl)methoxy-((((2R,3S,4R,5R)-5-(3-carbamoyl-4H-pyridin-1-yl)-3,4-dihydroxy-oxolan-2-yl)methoxy)hydroxyphosphoryl)oxyphosphinic acid disodium saltNicotinamide adenine dinucleotide reduced, disodium salt (reduced form)

N1209-1KGBL
700009-188EA 18939.53 USD
700009-188
β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na₂, reduced form), reduced
β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na2, reduced form)

Beta-Nicotinamide Adenine Dinucleotide Disodium Salt, Trihydrate, Reduced can be used in enzyme cycling assays to amplify detection of activity of biologically relevant enzymes or metabolites present in low concentrations.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₂₁H₂₇N₇Na₂O₁₄P₂
MW: 709.41 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00036200
CAS Number: 606-68-8
EINECS: 210-123-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR