2-Methyl-1,3-propanediol

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 1,3-Dihydroxy-2-methylpropane

M2610-3LT M2610-1LT M2610-200LTB
700002-980EA 264.8 USD
700002-980 700002-978 700002-976
2-Methyl-1,3-propanediol
2-Methyl-1,3-propanediol

2-Methyl-1,3-propanediol is used as a solvent in personal care products and also acts as a neutralizer, emollient, emulsifier, and humectant.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₄H₁₀O₂
MW: 90.12 g/mol
Density: 1.015
Storage Temperature: Ambient
MDL Number: MFCD00082586
CAS Number: 2163-42-0
EINECS: 412-350-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR