Methyl stearate

Synonyms: Methyl octadecanoate Stearic acid methyl ester

M2486-25GM M2486-100GM
700005-014EA 332.19 USD
700005-014 700005-016
Methyl stearate
Methyl stearate

Methyl Stearate is used as a nonionic surfactant in various experiments in helping solubilize a variety of chemical species by dissociating aggregates and unfolding proteins.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: CH₃(CH₂)₁₆CO₂CH₃
MW: 298.51 g/mol
Boiling Pt: 355 °C (1013 hPa)
Melting Pt: 37…39 °C
Density: 1.007 g/cm³ (20 °C)
Storage Temperature: Freezer
MDL Number: MFCD00009005
CAS Number: 112-61-8
EINECS: 203-990-4
Merck Index: 12,08959

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR