Methyl anthranilate ≥96% (DEA List I Chemical)
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
o-Aminobenzoic acid methyl ester
, Methyl 2-aminobenzoate
, Methyl 2-Anthranilate
, 2-Aminobenzoic acid methyl ester
M2340-250GM
700007-366EA
227
USD
700007-366
Methyl anthranilate ≥96% (DEA List I Chemical)
Methyl anthranilate
Formula:
C₈H₉NO₂ MW: 151.16 g/mol Boiling Pt: 237 °C (1013 hPa) Melting Pt: 24 °C Density: 1,16 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007710 CAS Number: 134-20-3 EINECS: 205-132-4 Merck Index: 12,06099 |
Specification Test Results
Assay | 96% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...