Hydroquinone monomethyl ether ≥98%
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Warning
|
Synonyms:
p-Methoxyphenol
, Mequinol
, 4-Methoxyphenol
, 1-Hydroxy-4-methoxybenzene
, 4-Hydroxyanisole
4-Methoxyphenol is a white, waxy solid with a phenolic, caramel odor. Also known as Mequinol and MeHQ, it is soluble in benzene, acetone, and alcohol. It is used as an antioxidant to inhibit the radical polymerization of monomers, a phenol in dermatology, and in dyestuffs. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for consumption.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₇H₈O₂ MW: 124.14 g/mol Boiling Pt: 243 °C (1013 hPa) Melting Pt: 55…57 °C Density: 1.55 g/cm³ (20 °C) Flash Pt: 128 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002332 CAS Number: 150-76-5 EINECS: 205-769-8 |
Specification Test Results
Assay | 98% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...