Polydimethylsiloxane , Melting Point Bath Oil

Synonyms: dimethiconePDMS

M1526-100ML
700012-834EA 173.96 USD
700012-834
Polydimethylsiloxane , Melting Point Bath Oil
Polydimethylsiloxane

Melting Point Bath Oil is a polydimethylsiloxane oil used for melting point and boiling point apparatuses with a temperature range of -40°C to +175°C.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: (CH₃)₃SiO(Si(CH₃)₂O)n
Density: 0.96
Storage Temperature: Ambient
MDL Number: MFCD00084411
CAS Number: 63148-62-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR