1-Tetradecanol ≥97%, Reagent Grade

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 1-Tetradecyl alcohol1-Hydroxy tetradecaneMyristyl alcohol

M1350-500GM M1350-100GM
700002-792EA 119.77 USD
700002-792 700002-794
1-Tetradecanol ≥97%, Reagent Grade
1-Tetradecanol

Myristyl Alcohol, Reagent, also known as Tetradecyl alcohol, is used as an ingredient in cosmetics such as cold creams for its emollient properties. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Formula: H₃C(CH₂)₁₂CH₂OH
MW: 214.39 g/mol
Boiling Pt: 289 °C (1013 hPa)
Melting Pt: 37…39 °C
Density: 0.823
Storage Temperature: Ambient
MDL Number: MFCD00004757
CAS Number: 112-72-1
EINECS: 204-000-3
Merck Index: 12,06418

Order Now

Specification Test Results

Assay 97% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR