Mercaptoacetic acid (thioglycolic acid) ≥98%, Reagent Grade

Danger

Synonyms: Mercaptoacetic acid

M1126-250ML
700008-120EA 190.99 USD
700008-120
Mercaptoacetic acid (thioglycolic acid) ≥98%, Reagent Grade
Mercaptoacetic acid (Thioglycolic acid)

Mercaptoacetic Acid, Reagent, also known as thioglycolic acid or TGA, is an organic compound with both carboxylic acid and thiol functional groups and is used as a chemical depilatory. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Formula: HSCH₂COOH
MW: 92.12 g/mol
Boiling Pt: 96 °C (1013 hPa)
Melting Pt: –16.5 °C
Density: 1.325
Flash Pt: 130 °C (closed cup)
Storage Temperature: Refrigerator
MDL Number: MFCD00004876
CAS Number: 68-11-1
EINECS: 200-677-4
UN: 1940
ADR: 8,II
Merck Index: 12,09472
https://us.vwr.com/stibo/web/std.lang.all/90/10/2299010.jpg

Order Now

Specification Test Results

Assay 98% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR