Manganese(II) sulfate monohydrate ≥98%, powder USP
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Warning
|
Synonyms:
Manganous sulfate monohydrate
, Manganous sulphate monohydrate
, Manganese sulfate monohydrate
, Manganese sulphate monohydrate
, Manganese(II) sulphate hydrate
, Manganese(II) sulfate hydrate
Manganese Sulfate, Monohydrate, Powder, USP is used as a trace element added to parenteral nutrition (PN) to prevent manganese deficiency; orally as a dietary supplement.
All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formula:
MnSO₄·1H₂O MW: 169.02 g/mol Melting Pt: 700 °C Density: 2.95 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149159 CAS Number: 10034-96-5 EINECS: 232-089-9 UN: 3077 ADR: 9,III Merck Index: 13,05763 |
Specification Test Results
Assay | 98% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...