Manganese(IV) oxide ≥99%, Reagent Grade

Supplier: SPECTRUM CHEMICAL MFG CORP

Danger

Synonyms: Manganese dioxideManganese(IV) oxide, activatedMangan(IV)-oxidManganese peroxide

M1107-12KG M1107-125GM M1107-500GM M1107-50KGBL M1107-2.5KG M1107-25GM
700000-840EA 2079.91 USD InStock
700000-840 700000-838 700000-846 700000-848 700000-842 700000-844
Manganese(IV) oxide ≥99%, Reagent Grade
Manganese (IV) oxide

Manganese Dioxide, Reagent is an inorganic compound that is black or brown in color. It is a component of dry cell batteries, as an inorganic pigment in ceramics and in glassmaking and as an oxidizing agent in organic synthesis. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Formula: MnO₂
MW: 86.94 g/mol
Melting Pt: 535 °C
Density: 5.03 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003463
CAS Number: 1313-13-9
EINECS: 215-202-6
Merck Index: 13,05753

Order Now

Specification Test Results

Assay 99% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR