Lithium fluoride ≥99%, powder, Reagent Grade

Warning

L1115-125GM L1115-2.5KG L1115-500GM L1115-12KGBL
700000-984EA 1127.27 USD
700000-984 700000-986 700000-980 700000-982
Lithium fluoride ≥99%, powder, Reagent Grade
Lithium fluoride

Lithium Fluoride, Powder, Reagent is most widely used as a flux in the production of ceramics, such as enamels, glasses and glazes. It is also used in brazing and welding fluxes and molten salt chemistry in metallurgy. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Formula: FLi
MW: 25.94 g/mol
Boiling Pt: 1676 °C (1013 hPa)
Melting Pt: 870 °C
Density: 2.64 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011090
CAS Number: 7789-24-4
EINECS: 232-152-0
UN: 3288
ADR: 6.1,III
Merck Index: 13,05553

Order Now

Specification Test Results

Assay 99% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR