Isatin ≥98%
Synonyms:
Indoline-2,3-dione
, 2,3-Indolinedione
, 1H-Indole-2,3-dione
IS105-25KGBL
IS105-500GM
IS105-100GM
700008-848EA
5947.96
USD
700008-848
700008-850
700008-846
Isatin ≥98%
Isatin
Isatin is used as a raw material in synthetic organic chemistry.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₈H₅NO₂ MW: 147.13 g/mol Boiling Pt: 342 °C (1013 hPa) Melting Pt: 198…202 °C Density: 1.472 g/cm³ (20 °C) Flash Pt: 220 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005718 CAS Number: 91-56-5 EINECS: 202-077-8 Merck Index: 13,05119 |
Specification Test Results
Assay | 98% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...