Indol-3-acetic acid ≥98%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 1H-Indole-3-acetic acidIAA3-Indoleacetic acidHeteroauxin

IN125-5KGBL
700009-160EA 10414 USD
700009-160
Indol-3-acetic acid ≥98%
Indol-3-acetic acid

Indole-3-acetic Acid is an aromatic heterocyclic organic compound that has a distinct bicyclic structure. It is a precursor to many pharmaceuticals and is a common component of fragrances and is a plant growth regulator. It is widely used in the production of Jasmine oil and is an auxin growth regulator, stimulates root growth and is plant tissue culture tested.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
Boiling Pt: 120…125 °C (0.05 mmHg)
Melting Pt: 167…170 °C
Storage Temperature: Ambient
MDL Number: MFCD00005636
CAS Number: 87-51-4
EINECS: 201-748-2
Merck Index: 13,04986

Order Now

Specification Test Results

Assay 98% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR