Triiodomethane ≥99% USP
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Warning
|
Synonyms:
Iodoform
I2031-25KGBL
700002-432EA
7160
USD
700002-432
Triiodomethane ≥99% USP
Triiodomethane
Iodoform, USP is a light yellow substance with a pungent odor and similar to chloroform and is an organoiodine compound. Iodoform has been used as a disinfectant. The USP grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formula:
CHI₃ MW: 393.73 g/mol Boiling Pt: 218 °C (1013 hPa) Melting Pt: 123 °C Density: 4.008 Storage Temperature: Ambient |
MDL Number:
MFCD00001069 CAS Number: 75-47-8 EINECS: 200-874-5 Merck Index: 13,05055 |
Specification Test Results
Assay | 99% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...