L(+)-Isoleucine ≥98.5% USP

Synonyms: (2S,3S)-(+)-IsoleucineH-Ile-OHL-Isoleucine

I1054-25KGBL I1054-5KGBL I1054-1KG I1054-100GM
700007-328EA 2402.84 USD
700007-328 95033-992 95033-994 700007-326
L(+)-Isoleucine ≥98.5% USP
L(+)-Isoleucine

L-Isoleucine, USP is one of nine essential amino acids in humans.


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: H₃CCH₂CH(CH₃)CH(NH₂)COOH
MW: 131.17 g/mol
Melting Pt: 279…280 °C
Storage Temperature: Ambient
MDL Number: MFCD00064222
CAS Number: 73-32-5
EINECS: 200-798-2
Merck Index: 13,05198

Order Now

Specification Test Results

Assay 98.5% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR