Isoamyl alcohol ≥98.5% ACS
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Danger
|
Synonyms:
3-Methyl-1-butanol
, Isopentyl alcohol
, 3-Methylbutanol
, iso-Pentanol
Isopentyl Alcohol, Reagent, ACS is also known as isoamyl alcohol and is a colorless, clear alcohol. It has an odor of banana oil. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.
Formula:
(CH₃)₂CHCH₂CH₂OH MW: 88.15 g/mol Boiling Pt: 130 °C (1013 hPa) Melting Pt: –117 °C Density: 0.809 Flash Pt: 42 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002934 CAS Number: 123-51-3 EINECS: 204-633-5 UN: 1105 ADR: 3,III Merck Index: 13,05214 |
Specification Test Results
Assay | 98.5% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...