DL-Histidine ≥99%

Synonyms: H-DL-His-OH2-Amino-3-(1H-imidazol-4-yl)propanoic acid

H3305-100GM H3305-25GM
700008-628EA 1046.84 USD
700008-628 700008-630
DL-Histidine ≥99%
DL-Histidine

DL-Histidine is a skin-conditioning amino acid.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₆H₉N₃O₂
MW: 155.16 g/mol
Melting Pt: 273 °C
Storage Temperature: Ambient
MDL Number: MFCD00005208
CAS Number: 4998-57-6
EINECS: 206-513-8
Merck Index: 12,04758

Order Now

Specification Test Results

Assay 99% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR