3-Phenylpropionic acid ≥99%
|
Warning
|
Synonyms:
3-Phenylpropanoic acid
, Hydrocinnamic acid
H2268-500GM
H2268-25GM
H2268-100GM
700005-706EA
116.84
USD
700005-706
700005-708
700005-710
3-Phenylpropionic acid ≥99%
3-Phenylpropionic acid
Hydrocinnamic Acid, a Phenylpropanoic acid, is a carboxylic acid used in cosmetics, pharmaceuticals, and food additives.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₉H₁₀O₂ MW: 150.18 g/mol Boiling Pt: 280 °C (1013 hPa) Melting Pt: 46…47 °C Density: 1.071 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002771 CAS Number: 501-52-0 EINECS: 207-924-5 Merck Index: 13,04805 |
Specification Test Results
Assay | 99% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...