2-Hydroxyethyl methacrylate ≥96% stabilized

Supplier: SPECTRUM CHEMICAL MFG CORP

Warning

Synonyms: HEMAMethacrylic acid 2-hydroxyethyl esterGMAGlycol methacrylate1,2-Ethanediol mono(2-methylpropenoate)

Stabilization: Stabilised with 4-Methoxyphenol (MEHQ) 250 ppm

H2262-500GM H2262-100GM
700005-402EA 138.67 USD
700005-402 700005-400
2-Hydroxyethyl methacrylate ≥96% stabilized
2-Hydroxyethyl methacrylate

2-Hydroxyethyl Methacrylate, Stabilized with 250 ppm MEHQ, also known as Hydroxyethyl methacrylate or HEMA, is used to make the polymer polyhydroxyethylmethacrylate which forms a hydrogel in water. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Formula: C₆H₁₀O₃
MW: 130.14 g/mol
Density: 1,073
Storage Temperature: Refrigerator
MDL Number: MFCD00002863
CAS Number: 868-77-9
EINECS: 212-782-2

Order Now

Specification Test Results

Assay 96% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR