Hydroxyurea ≥97% USP
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
N-(Aminocarbonyl)hydroxylamine
, Hydroxycarbamide
Hydroxyurea, USP is an antineoplastic drug used in myeloproliferative disorders. It has also been considered useful in treating painful attacks in sickle-cell disease and diseases like AIDS. The USP grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formula:
CH₄N₂O₂ MW: 76.05 g/mol Boiling Pt: 296 °C (1013 hPa) Melting Pt: 142…144 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007943 CAS Number: 127-07-1 EINECS: 204-821-7 Merck Index: 13,04874 |
Specification Test Results
Assay | 97% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...