HEPES-2-[4-(2-Hydroxyethyl)-1-piperazinyl]-ethanesulfonic acid ≥99%, Ultrapure biological buffer

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid2-(4-(2-Hydroxyethyl)piperazin-1-yl)ethanesulfonic acid2-(4-(2-Hydroxyethyl)piperazin-1-yl)ethanesulphonic acid4-(2-Hydroxyethyl)piperazin-1-ylethanesulfonic acidHEPES free acid

H1089-25GM H1089-5KGBL H1089-100GM H1089-250GM
700008-798EA 911 USD
700008-798 700008-796 700008-802 700008-800
HEPES-2-[4-(2-Hydroxyethyl)-1-piperazinyl]-ethanesulfonic acid ≥99%, Ultrapure biological buffer
HEPES (4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid)

HEPES, Biological Buffer, Ultrapure is as it's name suggests, a buffering agent. It's a zwitterionic organic chemical that is one of the '20 Good's buffers. Because it's better at maintaining physiological pH despite changes in carbon dioxide, it is widely used in cell culture. It has a decrease of water dissociation with falling temperature, this is especially relevant when working at low temperatures as it maintains enzyme function and structure. That said, it is important when working with HEPES for that particular purpose to keep the HEPES-containing solution in darkness as much as possible

Formula: C₈H₁₈N₂O₄S
MW: 238.31 g/mol
Melting Pt: 210…215 °C
Density: 1.448 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006158
CAS Number: 7365-45-9
EINECS: 230-907-9
Merck Index: 13,04672
https://us.vwr.com/stibo/web/std.lang.all/38/07/41813807.jpg

Order Now

Specification Test Results

Assay 99% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR