Hydrochloric acid 36.5% ACS
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Danger
|
Hydrochloric Acid, Reagent, ACS, is a colorless and clear, highly corrosive, highly pungent solution of hydrogen chloride in water. As an monoprotic acid and one of the six common strong mineral acids, it is the least likely to be subject to an oxidation-reduction reaction. The fact that it is available as a pure reagent make it an excellent acidifying reagent. Hydrochloric acid is also frequently used during chemical analysis to prepare samples for analysis. It reacts with basic compounds, turning them into dissolved chlorides that can then undergo analysis. As an ACS grade Reagent, Spectrum Chemical manufactured Hydrochloric Acid is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness
Formula:
HCl MW: 36.46 g/mol Boiling Pt: 110 °C (1013 hPa) Melting Pt: –30 °C Density: 1.200 Storage Temperature: Ambient |
MDL Number:
MFCD00011324 CAS Number: 7647-01-0 EINECS: 231-595-7 UN: 1789 ADR: 8,II |
Specification Test Results
Assay | 36.5% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...