1-Hexanol ≥98%, purified

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

Synonyms: Hexyl alcohol1-Hydroxy hexane1-Hexyl alcoholPentyl carbinol

H1012-55002 H1012-200LTB
700002-760EA 6292 USD
700002-760 CA11027-326
1-Hexanol ≥98%, purified
1-Hexanol

1-Hexanol, Purified is an organic alcohol that is a colorless liquid that is slightly soluble in water and most often used in the perfume industry. It is miscible with ethanol and ether. The Purified Grade describes chemicals of good quality where there are no official standards.

Formula: H₃C(CH₂)₄CH₂OH
MW: 102.18 g/mol
Boiling Pt: 156 °C (1013 hPa)
Melting Pt: –45 °C
Density: 0.814
Flash Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00002982
CAS Number: 111-27-3
EINECS: 203-852-3
UN: 2282
ADR: 3,III
Merck Index: 13,04715
https://us.vwr.com/stibo/web/std.lang.all/12/48/731248.jpg

Order Now

Specification Test Results

Assay 98% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR