Guanidinium carbonate ≥99%, Reagent Grade

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

Synonyms: Guanidine carbonateGuanidine carbonate saltDiguanidinium carbonate

G1043-1KG G1043-250GM G1043-5KG
700007-934EA 541.64 USD
700007-934 700007-930 700007-932
Guanidinium carbonate ≥99%, Reagent Grade
Guanidinium carbonate

Guanidine Carbonate, Reagent is sometimes used as a hair relaxer prior to use with thio-based hair straighteners. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Formula: C₃H₁₂N₆O₃
MW: 180.17 g/mol
Melting Pt: 197 °C
Density: 1.251 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013029
CAS Number: 593-85-1
EINECS: 209-813-7

Order Now

Specification Test Results

Assay 99% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR