Guanine ≥98%, Reagent Grade
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
2-Amino-1H-purin-6(7H)-one
G1029-25GM
G1029-1KG
G1029-5GM
G1029-100GM
700008-948EA
85
USD
700008-948
700008-942
700008-946
700008-944
Guanine ≥98%, Reagent Grade
Guanine
Guanine, Reagent, also known as 2-amino-6-hydroxypurine, is one of the four main nucleobases found in the nucleic acids DNA and RNA. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.
Formula:
C₅H₅N₅O MW: 151.13 g/mol Melting Pt: > 220 °C Density: 1.67 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00071533 CAS Number: 73-40-5 EINECS: 200-799-8 Merck Index: 13,04580 |
Specification Test Results
Assay | 98% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...