Ethyl octanoate ≥99%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Caprylic acid ethyl esterOctanoic acid ethyl esterEthyl caprylate

E2204-25ML E2204-100ML
700005-128EA 116.84 USD
700005-128 700005-130
Ethyl octanoate ≥99%
Ethyl octanoate

Ethyl Caprylate is a fatty acid ethyl ester and a octanoate ester.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₀H₂₀O₂
MW: 172.27 g/mol
Boiling Pt: 206 °C (1013 hPa)
Melting Pt: –47 °C
Density: 0.878
Flash Pt: 81 °C
Storage Temperature: Ambient
MDL Number: MFCD00009552
CAS Number: 106-32-1
EINECS: 203-385-5
Merck Index: 13,03813

Order Now

Specification Test Results

Assay 99% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR