Ethyl cinnamate
|
Warning
|
Synonyms:
3-Phenyl-acrylicacidethylester
, Cinnamic acid ethyl ester
Ethyl trans-Cinnamate, also known as Cinnamic Acid Ethyl Ester, is an organic compound and crystalline compound used in synthetic indigo, in flavors, and in certain pharmaceuticals.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₁₁H₁₂O₂ MW: 176.21 g/mol Boiling Pt: 271 °C (1013 hPa) Melting Pt: 7 °C Density: 1.049 Storage Temperature: Refrigerator |
MDL Number:
MFCD00009189 CAS Number: 103-36-6 EINECS: 203-104-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...