2-Ethoxyethanol, Reagent Grade
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Danger
|
Synonyms:
Cellosolve
, Ethylene glycol monoethyl ether
2-Ethoxyethanol, Reagent is a clear and colorless liquid that is water miscible and also referred to as ethyl cellosolve, a widely used solvent. It is used to dissolve chemically diverse compounds such as oils, resins, waxes among others. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.
Formula:
C₂H₅OCH₂CH₂OH MW: 90.12 g/mol Boiling Pt: 135 °C (1013 hPa) Melting Pt: –70 °C Density: 0.932 g/cm³ (20 °C) Flash Pt: 40 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00002869 CAS Number: 110-80-5 EINECS: 203-804-1 UN: 1171 ADR: 3,III REACH: 01-2119560582-38 Merck Index: 13,03786 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...