4-Hydroxy-4-methyl-2-pentanone
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Danger
|
Synonyms:
Diacetone alcohol
DI102-500MLG
DI102-200LTB
DI102-4LTGL
DI102-20LTMT
700004-060EA
122
USD
700004-060
700004-054
700004-058
700004-056
4-Hydroxy-4-methyl-2-pentanone
4-Hydroxy-4-methyl-2-pentanone
Diacetone Alcohol is a chemical compound that is liquid and used as a synthetic intermediate in the preparation of other compounds such as cellulose ester lacquers, lacquer thinners and wood stains.
Formula:
(CH₃)₂C(OH)CH₂COCH₃ MW: 116.16 g/mol Boiling Pt: 166 °C (1013 hPa) Melting Pt: –44 °C Density: 0.94 g/cm³ (20 °C) Flash Pt: 58 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004471 CAS Number: 123-42-2 EINECS: 204-626-7 UN: 1148 ADR: 3,III REACH: 01-2119473975-21 Merck Index: 13,02983 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...