Laurinaldehyde ≥92%

Synonyms: Lauric aldehydeDodecanalDodecylaldehyde

D3837-100GM
700003-696EA 122.03 USD
700003-696
Laurinaldehyde ≥92%
Laurinaldehyde
Formula: C₁₂H₂₄O
MW: 184.32 g/mol
Boiling Pt: 242 °C (1013 hPa)
Melting Pt: 44 °C
Density: 0.835
Storage Temperature: Ambient
MDL Number: MFCD00007017
CAS Number: 112-54-9
EINECS: 203-983-6
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Assay 92% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR