Dodecyltrimethylammonium bromide
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Dodecyl trimethyl ammonium bromide
, (1-Dodecyl)trimethylammonium bromide
D3009-25GM
D3009-100GM
D3009-5GM
700007-614EA
85
USD
700007-614
700007-610
700007-612
Dodecyltrimethylammonium bromide
Dodecyltrimethylammonium bromide
Dodecyltrimethylammonium Bromide is a quaternary ammonium salt useful as paint strippers, foaming stabilizers, and bactericidal lotions.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₁₅H₃₄BrN MW: 308.35 g/mol Melting Pt: 264…266 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011767 CAS Number: 1119-94-4 EINECS: 214-290-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...