Diethyl succinate ≥99%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Succinic acid diethyl ester1,4-Butanedioic acid diethyl ester

D2138-200LTB D2138-500GM
700005-868EA 9967 USD
700005-868 700005-870
Diethyl succinate ≥99%
Diethyl succinate

Diethyl Succinate, also known as Butanedioic acid diethyl ester, is often used in fragrances.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₈H₁₄O₄
MW: 174.2 g/mol
Boiling Pt: 217 °C (1013 hPa)
Melting Pt: –19 °C
Density: 1.047
Flash Pt: 80 °C
Storage Temperature: Ambient
MDL Number: MFCD00009208
CAS Number: 123-25-1
EINECS: 204-612-0
Merck Index: 12,09038

Order Now

Specification Test Results

Assay 99% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR