Dehydroacetic acid ≥98%

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

Synonyms: 3-Acetyl-6-methyl-2H-pyran-2,4(3H)-dione3-Acetyl-4-hydroxy-6-methyl-2H-pyran-2-one

D2012-500GM D2012-100GM
700008-350EA 181.14 USD
700008-350 700008-348
Dehydroacetic acid ≥98%
Dehydracetic acid

Dehydroacetic Acid, also known as Methylacetopyronone, is an organic compound used as a plasticizer in synthetic resins, a preservative in foods and as an antienzyme in toothpastes.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₈H₈O₄
MW: 168.15 g/mol
Boiling Pt: 333 °C (1013 hPa)
Melting Pt: 108 °C
Flash Pt: 157 °C
Storage Temperature: Ambient
MDL Number: MFCD00066709
CAS Number: 520-45-6
EINECS: 208-293-9
Merck Index: 13,02885

Order Now

Specification Test Results

Assay 98% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR