Sudan III

Synonyms: Sudan Red IIID and C Red No. 17Sudan Red BK1-(4-(Phenylazo)phenylazo)-2-naphtholSudan® III (Red)Sudan GCerasin RedTony RedOil scarletScarlet BOrganol® red BSFat Ponceau GOil redSolvent Red 23Sudan Red B1-((4-(Phenyldiazenyl)phenyl)diazenyl)naphthalen-2-olSudan V

D1132-500GM D1132-25GM D1132-5KGBL
700010-270EA 1413.44 USD
700010-270 700010-268 700010-272
Sudan III
Sudan III

D and C Red No. 17 | Non-graded D and C Red No. 17 supplied by Spectrum is indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₂₂H₁₆N₄O
MW: 352.4 g/mol
Melting Pt: 205.3…206.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00003905
CAS Number: 85-86-9
EINECS: 201-638-4
Merck Index: 12,09054

Order Now

Specification Test Results

Assay 90% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR