DL-Camphor (synthetic), block

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

Synonyms: DL-1,7,7-Trimethylbicyclo[2.2.1]-2-heptanone(±)-1,7,7-Trimethyl-2-norbornanone1,7,7-Trimethylbicyclo[2.2.1]heptan-2-one(±)-1,7,7-Trimethylbicyclo[2.2.1]-2-heptanone

CA186-1LB
700003-996EA 259 USD
700003-996
DL-Camphor (synthetic), block
DL-Camphor

Camphor, Synthetic, Blocks is a white to transparent solid that has a waxy texture, strong aromatic odor and flammable. It is found naturally in the wood of the camphor laurel as well as in dried rosemary, mint and is a terpinoid. It has various uses such as embalming fluid, religious ceremonies, in cooking, for it's scent as well as for medicinal purposes in products like Vick's Vaporub.

Formula: C₁₀H₁₆O
MW: 152.24 g/mol
Boiling Pt: 204 °C (1013 hPa)
Melting Pt: ∼180 °C
Density: 0.992
Flash Pt: 66 °C
Storage Temperature: Ambient
MDL Number: MFCD00074738
CAS Number: 76-22-2
EINECS: 200-945-0
UN: 2717
ADR: 4.1,III

Order Now

Specification Test Results

Assay 96% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR