Pentadecan-15-olide
Synonyms:
Oxacyclohexadecan-2-one
, omega-Pentadecalactone
, ω-Pentadecalactone
, Oxa-2-cyclohexadecanone
, Cyclopentadecanolide
, Pentadecanolide
Cyclopentadecanolide, also known as Angelica lactone or Muskalactone, can be used as a flavoring agent.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₁₅H₂₈O₂ MW: 240.39 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00039667 CAS Number: 106-02-5 EINECS: 203-354-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...