Calcium hydrogen orthophosphate, anhydrous ≥98%, Powder USP

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Calcium phosphate dibasicAnhydrous Dibasic Calcium PhosphateCalcium hydrogen phosphateCalcium perphosphate, ammonifiedCalcium phosphate, dibasic

C1890-500GM C1890-2.5KG C1890-12KG
700000-368EA 553 USD
700000-368 700000-370 700000-366
Calcium hydrogen orthophosphate, anhydrous ≥98%, Powder USP
Calcium hydrogen orthophosphate

Anhydrous Dibasic Calcium Phosphate, Powder, USP is used as a dietary supplement and as an antacid. It is listed on the FDA's Generally Recognized As Safe (GRAS) list of substances. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: CaHPO₄
MW: 136.06 g/mol
Melting Pt: 370 °C
Density: 2.89 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010909
CAS Number: 7757-93-9
EINECS: 231-826-1
Merck Index: 12,01739

Order Now

Specification Test Results

Assay 98% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR