Cyclohexane ≥99% ACS
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Danger
|
Synonyms:
Hexahydro-Benzene
, Hexamethylene
C1455-100ML
C1455-1LT
C1455-200LTB
C1455-500MLG
C1455-20LTMT
C1455-4LTGL
700002-330EA
299
USD
700002-330
700002-328
700002-326
700002-324
700002-332
700002-322
Cyclohexane ≥99% ACS
Cyclohexane
Cyclohexane, Reagent, ACS is used as a nonpolar solvent, in the calibration of differential scanning calorimetry and other industrial applications including a chemical intermediate. The distinctive detergent-like odor of cyclohexane is reminiscent of cleaning product and is sometimes used in cleaning products as a fragrance enhancer
Formula:
C₆H₁₂ MW: 84.16 g/mol Boiling Pt: 81 °C (1013 hPa) Melting Pt: 7 °C Density: 0.779 Flash Pt: –18 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003814 CAS Number: 110-82-7 EINECS: 203-806-2 UN: 1145 ADR: 3,II Merck Index: 13,02752 |
Specification Test Results
Assay | 99% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...