Calcium peroxide ≥60% FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

C1388-25GM C1388-500GM C1388-125GM
95030-838EA 140 USD
95030-838 95030-842 95030-840
Calcium peroxide ≥60% FCC
Calcium peroxide

Calcium Peroxide, FCC is used as a food additive under the E number E930 as flour bleaching agent. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: CaO₂
Melting Pt: 692 °C
Density: 2.919…2.921 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00015988
CAS Number: 1305-79-9
EINECS: 215-139-4
UN: 1457
ADR: 5.1,II
Merck Index: 12,01736

Order Now

Specification Test Results

Assay 60% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR